Assay Details
Gene Symbol: DL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340127.1 | NP_001325985.1 | 12 - 13 | 1940 | 96 | |
NM_116041.3 | NP_191735.2 | 13 - 14 | 1846 | 96 | ||
GenBank mRNA | AY189279.1 | - | 13 - 14 | 1770 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
