Assay Details
| Gene Symbol: RS31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116051.4 | NP_567120.1 | 5 - 6 | 914 | 68 | |
| GenBank mRNA | AF439831.1 | - | 5 - 6 | 853 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RS31| Gene Symbol: RS31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116051.4 | NP_567120.1 | 5 - 6 | 914 | 68 | |
| GenBank mRNA | AF439831.1 | - | 5 - 6 | 853 | 68 | |
Set Membership: |
|