Assay Details
| Gene Symbol: EDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116171.3 | NP_567137.1 | 1 | 1433 | 140 | |
| GenBank mRNA | AY056271.1 | - | 1 | 1041 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDL3| Gene Symbol: EDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116171.3 | NP_567137.1 | 1 | 1433 | 140 | |
| GenBank mRNA | AY056271.1 | - | 1 | 1041 | 140 | |
Set Membership: |
|