Assay Details
| Gene Symbol: EMB3126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116214.3 | NP_191908.1 | 6 - 7 | 968 | 109 | |
| NM_202757.2 | NP_974486.1 | 6 - 7 | 958 | 109 | ||
| GenBank mRNA | AY053410.1 | - | 6 - 7 | 956 | 109 | |
| AY059929.1 | - | 6 - 7 | 955 | 109 | ||
| AY133519.1 | - | 6 - 7 | 902 | 109 | ||
| BT001191.1 | - | 6 - 7 | 902 | 109 | ||
More Information
Set Membership: |
|

EMB3126
Genomic Map