Assay Details
| Gene Symbol: RHB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340240.1 | NP_001319828.1 | 3 - 4 | 687 | 81 | |
| NM_116256.4 | NP_567171.1 | 3 - 4 | 669 | 81 | ||
| NM_202759.2 | NP_974488.1 | 3 - 4 | 622 | 81 | ||
| GenBank mRNA | AK228408.1 | - | 3 - 4 | 628 | 81 | |
| BT024583.1 | - | 2 - 3 | 425 | 81 | ||
More Information
Set Membership: |
|

RHB1A
Genomic Map