Assay Details
Gene Symbol: ROPGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125431.2 | NP_001118903.1 | 5 - 6 | 922 | 118 | |
NM_001340252.1 | NP_001328184.1 | 3 - 4 | 918 | 118 | ||
NM_001340253.1 | NP_001328183.1 | 4 - 5 | 850 | 118 | ||
NM_116271.3 | NP_191956.2 | 5 - 6 | 922 | 118 | ||
GenBank mRNA | BT030385.1 | - | 4 - 5 | 659 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
