Assay Details
| Gene Symbol: MYB3R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116278.1 | NP_567179.1 | 8 - 9 | 1434 | 65 | |
| GenBank mRNA | AF151647.1 | - | 8 - 9 | 1208 | 65 | |
| AF218054.2 | - | 8 - 9 | 1434 | 65 | ||
More Information
Set Membership: |
|

MYB3R2
Genomic Map