Assay Details
| Gene Symbol: NAD-ME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116281.4 | NP_191966.2 | 15 - 16 | 1660 | 74 | |
| GenBank mRNA | AY128396.1 | - | 15 - 16 | 1576 | 74 | |
| BT000075.1 | - | 14 - 15 | 1500 | 74 | ||
| BT002046.1 | - | 15 - 16 | 1590 | 74 | ||
| BT008375.1 | - | 14 - 15 | 1500 | 74 | ||
More Information
Set Membership: |
|

NAD-ME2
Genomic Map