Assay Details
Gene Symbol: BSK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340265.1 | NP_001329175.1 | 9 - 10 | 2144 | 65 | |
NM_116296.4 | NP_191980.2 | 10 - 11 | 2156 | 65 | ||
GenBank mRNA | AY062495.1 | - | 10 - 11 | 1899 | 65 | |
AY093275.1 | - | 9 - 10 | 1219 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
