Assay Details
Gene Symbol: BSK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340265.1 | NP_001329175.1 | 1 - 2 | 999 | 87 | |
NM_116296.4 | NP_191980.2 | 2 - 3 | 1011 | 87 | ||
GenBank mRNA | AY062495.1 | - | 2 - 3 | 754 | 87 | |
AY093275.1 | - | 1 - 2 | 74 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
