Assay Details
| Gene Symbol: BSK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340265.1 | NP_001329175.1 | 8 - 9 | 2030 | 85 | |
| NM_116296.4 | NP_191980.2 | 9 - 10 | 2042 | 85 | ||
| GenBank mRNA | AY062495.1 | - | 9 - 10 | 1785 | 85 | |
| AY093275.1 | - | 8 - 9 | 1105 | 85 | ||
More Information
Set Membership: |
|

BSK3
Genomic Map