Assay Details
| Gene Symbol: AT4G00780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116304.4 | NP_567187.1 | 5 - 6 | 956 | 74 | |
| GenBank mRNA | AY065285.1 | - | 5 - 6 | 795 | 74 | |
| AY091337.1 | - | 5 - 6 | 750 | 74 | ||
More Information
Set Membership: |
|

AT4G00780
Genomic Map