Assay Details
Gene Symbol: ECA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340276.1 | NP_001329712.1 | 2 - 3 | 1841 | 72 | |
NM_116317.4 | NP_191999.1 | 3 - 4 | 2016 | 72 | ||
GenBank mRNA | AJ132387.1 | - | 2 - 3 | 1650 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
