Assay Details
| Gene Symbol: AT4G01340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116364.1 | NP_192043.1 | 1 - 2 | 101 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G01340| Gene Symbol: AT4G01340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116364.1 | NP_192043.1 | 1 - 2 | 101 | 112 | |
Set Membership: |
|