Assay Details
Gene Symbol: AT4G01340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116364.1 | NP_192043.1 | 3 - 4 | 350 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
