Assay Details
| Gene Symbol: CBL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340311.1 | NP_001329076.1 | 1 - 2 | 83 | 80 | |
| NM_116372.3 | NP_192051.2 | 1 - 2 | 83 | 80 | ||
| GenBank mRNA | AF192885.2 | - | 1 - 2 | 80 | 80 | |
| AF290435.1 | - | 1 - 2 | 80 | 80 | ||
More Information
Set Membership: |
|

CBL5
Genomic Map