Assay Details
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203726.1 | NP_001190655.1 | 4 - 5 | 261 | 87 | |
NM_116437.3 | NP_192115.1 | 2 - 3 | 211 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
