Assay Details
| Gene Symbol: MSH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203727.2 | NP_001190656.1 | 11 - 12 | 2585 | 65 | |
| NM_116438.2 | NP_192116.1 | 11 - 12 | 2595 | 65 | ||
More Information
Set Membership: |
|

MSH6
Genomic Map