Assay Details
| Gene Symbol: MOS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340367.1 | NP_001328074.1 | 8 - 9 | 1190 | 101 | |
| NM_116447.3 | NP_192124.1 | 8 - 9 | 1190 | 101 | ||
| GenBank mRNA | AF385693.1 | - | 8 - 9 | 1189 | 101 | |
| AY081725.1 | - | 8 - 9 | 1102 | 101 | ||
| DQ087970.1 | - | 8 - 9 | 1102 | 101 | ||
More Information
Set Membership: |
|

MOS6
Genomic Map