Assay Details
| Gene Symbol: MOS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116447.3 | NP_192124.1 | 9 - 10 | 1321 | 79 | |
| GenBank mRNA | AF385693.1 | - | 9 - 10 | 1320 | 79 | |
| AY081725.1 | - | 9 - 10 | 1233 | 79 | ||
| DQ087970.1 | - | 9 - 10 | 1233 | 79 | ||
More Information
Set Membership: |
|

MOS6
Genomic Map