Assay Details
Gene Symbol: SYP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116452.4 | NP_567223.1 | 5 - 6 | 868 | 102 | |
GenBank mRNA | AF154574.1 | - | 5 - 6 | 718 | 102 | |
AK229319.1 | - | 5 - 6 | 830 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
