Assay Details
Gene Symbol: RSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340369.1 | NP_001329795.1 | 4 - 5 | 779 | 111 | |
NM_116459.4 | NP_567226.1 | 4 - 5 | 796 | 111 | ||
NM_178956.4 | NP_849287.2 | 4 - 5 | 787 | 111 | ||
NM_202772.2 | NP_974501.1 | 4 - 5 | 787 | 111 | ||
GenBank mRNA | AF225702.1 | - | 4 - 5 | 640 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
