Assay Details
Gene Symbol: MLO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036501.2 | NP_001031578.1 | 2 - 3 | 350 | 74 | |
NM_116494.5 | NP_192169.1 | 1 - 2 | 434 | 74 | ||
GenBank mRNA | AY072135.1 | - | 1 - 2 | 281 | 74 | |
AY113992.1 | - | 1 - 2 | 181 | 74 | ||
Z95352.1 | - | 273 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
