Assay Details
Gene Symbol: EOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116502.4 | NP_192177.1 | 3 - 4 | 2816 | 68 | |
GenBank mRNA | AK229195.1 | - | 3 - 4 | 2734 | 68 | |
AY572792.1 | - | 3 - 4 | 2132 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
