Assay Details
| Gene Symbol: AT4G02940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116528.6 | NP_192203.1 | 2 - 3 | 1051 | 126 | |
| GenBank mRNA | AY093043.1 | - | 2 - 3 | 861 | 126 | |
| AY128924.1 | - | 2 - 3 | 706 | 126 | ||
More Information
Set Membership: |
|

AT4G02940
Genomic Map