Assay Details
| Gene Symbol: AOP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340416.1 | NP_001319854.1 | 3 - 4 | 1057 | 126 | |
| NM_001340417.1 | NP_001319855.1 | 3 - 4 | 1061 | 126 | ||
More Information
Set Membership: |
|

AOP3
Genomic Map