Assay Details
| Gene Symbol: AOP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116541.4 | NP_192216.1 | 1 - 2 | 560 | 86 | |
| GenBank mRNA | AF417855.1 | - | 1 - 2 | 385 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AOP1| Gene Symbol: AOP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116541.4 | NP_192216.1 | 1 - 2 | 560 | 86 | |
| GenBank mRNA | AF417855.1 | - | 1 - 2 | 385 | 86 | |
Set Membership: |
|