Assay Details
| Gene Symbol: PMP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340483.1 | NP_001319865.1 | 7 - 8 | 1298 | 66 | |
| NM_116685.5 | NP_192356.1 | 6 - 7 | 560 | 66 | ||
| GenBank mRNA | AJ006053.1 | - | 6 - 7 | 509 | 66 | |
| AK227405.1 | - | 6 - 7 | 529 | 66 | ||
| BT003055.1 | - | 6 - 7 | 506 | 66 | ||
More Information
Set Membership: |
|

PMP22
Genomic Map