Assay Details
Gene Symbol: RIC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340518.1 | NP_001319870.1 | 2 - 3 | 540 | 114 | |
NM_116728.2 | NP_192399.1 | 1 - 2 | 94 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
