Assay Details
Gene Symbol: SFR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203747.1 | NP_001190676.1 | 14 - 15 | 3567 | 64 | |
NM_001340519.1 | NP_001329057.1 | 14 - 15 | 3616 | 64 | ||
NM_116730.5 | NP_192401.5 | 14 - 15 | 3313 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
