Assay Details
Gene Symbol: SFR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203747.1 | NP_001190676.1 | 1 - 2 | 625 | 75 | |
NM_001340519.1 | NP_001329057.1 | 1 - 2 | 671 | 75 | ||
NM_116730.5 | NP_192401.5 | 1 - 2 | 338 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
