Assay Details
Gene Symbol: SFR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203747.1 | NP_001190676.1 | 7 - 8 | 1681 | 67 | |
NM_001340519.1 | NP_001329057.1 | 7 - 8 | 1727 | 67 | ||
NM_116730.5 | NP_192401.5 | 7 - 8 | 1394 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
