Assay Details
| Gene Symbol: CRK25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340543.1 | NP_001329426.1 | 6 - 7 | 1768 | 131 | |
| NM_116759.3 | NP_192429.1 | 6 - 7 | 1703 | 131 | ||
More Information
Set Membership: |
|

CRK25
Genomic Map