Assay Details
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116951.2 | NP_192621.1 | 1 | 1866 | 116 | |
GenBank mRNA | AY056392.1 | - | 1 | 1742 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
