Assay Details
| Gene Symbol: PUM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116954.2 | NP_192624.2 | 2 - 3 | 1158 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PUM11| Gene Symbol: PUM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116954.2 | NP_192624.2 | 2 - 3 | 1158 | 140 | |
Set Membership: |
|