Assay Details
| Gene Symbol: AT5G19730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121978.4 | NP_197474.1 | 2 - 3 | 848 | 95 | |
| GenBank mRNA | AY065431.1 | - | 2 - 3 | 665 | 95 | |
| AY096559.1 | - | 2 - 3 | 561 | 95 | ||
More Information
Set Membership: |
|

AT5G19730
Genomic Map