Assay Details
| Gene Symbol: AT5G19730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121978.4 | NP_197474.1 | 3 - 4 | 1060 | 81 | |
| GenBank mRNA | AY065431.1 | - | 3 - 4 | 877 | 81 | |
| AY096559.1 | - | 3 - 4 | 773 | 81 | ||
More Information
Set Membership: |
|

AT5G19730
Genomic Map