Assay Details
| Gene Symbol: AT5G19890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121996.3 | NP_568385.1 | 3 - 4 | 663 | 62 | |
| GenBank mRNA | AY123985.1 | - | 3 - 4 | 617 | 62 | |
| BT000582.1 | - | 3 - 4 | 574 | 62 | ||
More Information
Set Membership: |
|

AT5G19890
Genomic Map