Assay Details
| Gene Symbol: ROPGEF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104103.6 | NP_175634.6 | 2 - 3 | 614 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ROPGEF11| Gene Symbol: ROPGEF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104103.6 | NP_175634.6 | 2 - 3 | 614 | 72 | |
Set Membership: |
|