Assay Details
| Gene Symbol: BGLU18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198275.1 | NP_001185204.1 | 11 - 12 | 1487 | 103 | |
| NM_104118.3 | NP_175649.1 | 11 - 12 | 1641 | 103 | ||
| GenBank mRNA | AY039855.1 | - | 11 - 12 | 1456 | 103 | |
| AY056415.1 | - | 11 - 12 | 1456 | 103 | ||
| BT000515.1 | - | 11 - 12 | 1380 | 103 | ||
More Information
Set Membership: |
|

BGLU18
Genomic Map