Assay Details
| Gene Symbol: TSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104119.3 | NP_564607.1 | 23 - 24 | 1404 | 152 | |
| NM_179466.3 | NP_849797.1 | 23 - 24 | 1541 | 152 | ||
More Information
Set Membership: |
|

TSA1
Genomic Map