Assay Details
| Gene Symbol: TSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104119.3 | NP_564607.1 | 4 - 5 | 301 | 136 | |
| NM_179466.3 | NP_849797.1 | 4 - 5 | 426 | 136 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSA1| Gene Symbol: TSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104119.3 | NP_564607.1 | 4 - 5 | 301 | 136 | |
| NM_179466.3 | NP_849797.1 | 4 - 5 | 426 | 136 | ||
Set Membership: |
|