Assay Details
| Gene Symbol: MMH-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333547.1 | NP_001322993.1 | 1 - 2 | 390 | 74 | |
| NM_001333548.1 | NP_001322994.1 | 1 - 2 | 349 | 74 | ||
| NM_001333549.1 | NP_001322995.1 | 1 - 2 | 349 | 74 | ||
| NM_104128.3 | NP_564608.1 | 1 - 2 | 390 | 74 | ||
| NM_179467.3 | NP_849798.1 | 1 - 2 | 302 | 74 | ||
| GenBank mRNA | AF099971.1 | - | 1 - 2 | 277 | 74 | |
| BT043496.1 | - | 1 - 2 | 227 | 74 | ||
More Information
Set Membership: |
|

MMH-1
Genomic Map