Assay Details
Gene Symbol: NAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104166.3 | NP_175696.1 | 2 - 3 | 657 | 169 | |
GenBank mRNA | AF123311.1 | - | 2 - 3 | 542 | 169 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
