Assay Details
| Gene Symbol: CID8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036105.1 | NP_001031182.1 | 3 - 4 | 642 | 60 | |
| NM_104243.4 | NP_175769.1 | 3 - 4 | 832 | 60 | ||
| GenBank mRNA | BT014885.1 | - | 3 - 4 | 456 | 60 | |
More Information
Set Membership: |
|

CID8
Genomic Map