Assay Details
| Gene Symbol: MVP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333627.1 | NP_001323162.1 | 1 - 2 | 392 | 84 | |
| NM_104280.4 | NP_175805.1 | 1 - 2 | 392 | 84 | ||
| GenBank mRNA | BT010163.1 | - | 1 - 2 | 304 | 84 | |
More Information
Set Membership: |
|

MVP1
Genomic Map