Assay Details
| Gene Symbol: SPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104293.4 | NP_564653.1 | 5 - 6 | 919 | 73 | |
| GenBank mRNA | AY058075.1 | - | 5 - 6 | 917 | 73 | |
| AY143876.1 | - | 5 - 6 | 871 | 73 | ||
More Information
Set Membership: |
|

SPL2
Genomic Map