Assay Details
Gene Symbol: NF-YA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104294.3 | NP_175818.1 | 2 - 3 | 755 | 74 | |
GenBank mRNA | AF386957.1 | - | 2 - 3 | 664 | 74 | |
AY081445.1 | - | 1 - 2 | 465 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
