Assay Details
Gene Symbol: TAF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036110.1 | NP_001031187.2 | 11 - 12 | 1196 | 91 | |
NM_001198292.1 | NP_001185221.1 | 11 - 12 | 1131 | 91 | ||
NM_001333642.1 | NP_001319224.1 | 10 - 11 | 1283 | 91 | ||
NM_001333643.1 | NP_001322136.1 | 10 - 11 | 1327 | 91 | ||
NM_104314.6 | NP_175838.4 | 11 - 12 | 1259 | 91 | ||
NM_202295.2 | NP_974024.2 | 10 - 11 | 1312 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
