Assay Details
Gene Symbol: TAF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036110.1 | NP_001031187.2 | 6 - 7 | 598 | 95 | |
NM_001198292.1 | NP_001185221.1 | 6 - 7 | 533 | 95 | ||
NM_001333642.1 | NP_001319224.1 | 5 - 6 | 685 | 95 | ||
NM_001333643.1 | NP_001322136.1 | 5 - 6 | 729 | 95 | ||
NM_104314.6 | NP_175838.4 | 6 - 7 | 661 | 95 | ||
NM_202295.2 | NP_974024.2 | 5 - 6 | 714 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
